missing translation for 'onlineSavingsMsg'
Learn More

UBE2L6 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17278-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877144

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L6 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to function in the E6/E6-ap-induced ubiquitination of p53.
Specifications

Specifications

UBE2L6
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14933
RIG B, UBCH8, UBE2L6, Ubiquitin carrier protein L6, Ubiquitin protein ligase L6
Rabbit
IgG
150 μL
-20°C
Primary
9246
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
UBE2L6
Liquid
UBE2L6
UBE2L6 Fusion Protein Ag11045
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers