missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2W Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567547
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Rat samples.
UBE2W accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins, and catalyzes monoubiquitination. Further, UBE2W is involved in degradation of misfolded chaperone substrates by mediating monoubiquitination of STUB1/CHIP, leading to recruitment of ATXN3 to monoubiquitinated STUB1/CHIP, and restriction of the length of ubiquitin chain attached to STUB1/CHIP substrates by ATXN3. After UV irradiation, but not after mitomycin-C (MMC) treatment, acts as a specific E2 ubiquitin-conjugating enzyme for the Fanconi anemia complex by associating with E3 ubiquitin-protein ligase FANCL and catalyzing monoubiquitination of FANCD2, a key step in the DNA damage pathway. UBE2W In vitro catalyzes 'Lys-11'-linked polyubiquitination, and transfers ubiquitin in complex with RING/U-box type E3s that do not have active site cysteine residues to form thioester bonds with ubiquitin, and preferentially ubiquitinates the N-terminus of substrates, such as ATXN3, STUB1 and SUMO2.Specifications
UBE2W | |
Polyclonal | |
Unconjugated | |
UBE2W | |
6130401J04Rik; E2 ubiquitin-conjugating enzyme W; N-terminal E2 ubiquitin-conjugating enzyme; N-terminus-conjugating E2; probable ubiquitin-conjugating enzyme E2 W; UBC16; UBC-16; UBE2W; Ubiquitin carrier protein W; ubiquitin conjugating enzyme E2 W (putative); ubiquitin conjugating enzyme E2W (putative); ubiquitin-conjugating enzyme 16; ubiquitin-conjugating enzyme 2W; ubiquitin-conjugating enzyme E2 W; ubiquitin-conjugating enzyme E2 W-like; ubiquitin-conjugating enzyme E2W; ubiquitin-conjugating enzyme E2W (putative); ubiquitin-protein ligase W | |
Rabbit | |
affinity chromatography | |
RUO | |
100909445, 55284 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
B5DEI4, Q96B02 | |
UBE2W | |
Full length Fusion protein of human UBE2W | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |