Learn More
UBE3D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555552
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SLVIESLRNS KYIKKFPLLE NTFKADSSSA WSAVKVLYQP CIKSRNEKLV SLWESDISVH PLTLPSATCL ELLL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.
UBE3D Polyclonal antibody specifically detects UBE3D in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
UBE3D | |
Polyclonal | |
Unconjugated | |
UBE3D | |
C6orf157; E3 ubiquitin-protein ligase E3D; H10BH; HECT-type E3 ubiquitin transferase E3D; UBCH10 binding protein with a hect-like domain; ubcH10-binding protein with a HECT-like domain; UBE2CBP; UBE3D; ubiquitin protein ligase E3D; ubiquitin-conjugating enzyme E2C binding protein; ubiquitin-conjugating enzyme E2C-binding protein; YJR141W | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90025 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6J8 | |
UBE3D | |
Recombinant protein corresponding to Human UBE3D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only