missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18148-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.Specifications
UBE4B | |
Polyclonal | |
Unconjugated | |
UBE4B | |
E4, HDNB1, KIAA0684, UBE4B, UBOX3, UFD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10277, 298652, 63958 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95155, Q9ES00 | |
Ube4b | |
UBE4B Fusion Protein Ag12836 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |