Learn More
Ubinuclein 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564392
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AKKKVMAPSK IKVKESSTKP DKKVSVPSGQ IGGPIALPSD HQTGGLSIGA SSRELPSQAS GGLANPPPVN LEDSLDEDLI RNPASSVEA Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
Ubinuclein 1 Polyclonal antibody specifically detects Ubinuclein 1 in Human samples. It is validated for Immunohistochemistry.Specifications
Ubinuclein 1 | |
Polyclonal | |
Unconjugated | |
UBN1 | |
1110029L11Rik; 2610108L02Rik; AA673476; AW124741; HIRA-binding protein; Protein VT4; ubinuclein 1; ubinuclein-1; Ubiquitously expressed nuclear protein; Ubn1; VT; VT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPG3 | |
UBN1 | |
Recombinant protein corresponding to Human Ubinuclein 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only