missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550404
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total UCC1 protein.
UCC1 Polyclonal antibody specifically detects UCC1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
UCC1 | |
Polyclonal | |
Unconjugated | |
EPDR1 | |
AU040950; AV006210; Epdm2; EPDR; Epdr1; Epdr2; ependymin 2; ependymin related 1; ependymin related protein 1; ependymin related protein 1 (zebrafish); ependymin related protein 2; mammalian ependymin related protein 1; mammalian ependymin-related protein 1; MERP1; MERP-1; Merp2; MERP-2; Ucc1; upregulated in colorectal cancer gene 1; upregulated in colorectal cancer gene 1 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
105298, 291180, 54749 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q5XII0, Q99M71, Q9UM22 | |
EPDR1 | |
Fusion protein corresponding to residues near the C terminal of human ependymin related 1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |