missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
UCH-L1/PGP9.5 Antibody (BH7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-58869-0.025ML
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Specifikationer
| UCH-L1/PGP9.5 | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:20000, Simple Western 1:20000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 | |
| P09936 | |
| UCHL1 | |
| Recombinant full length human UCHL1 purified from E. coli. [Swiss-Prot# P09936] | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Bovine, Mouse, Equine, Pig | |
| Ascites |
| Immunohistochemistry (Free Floating), Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| BH7 | |
| Unconjugated | |
| 50% PBS, 50% Glycerol with 5mM Sodium Azide | |
| EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
| Mouse | |
| 24 kDa | |
| 0.025 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 7345 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering