missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCHL1/PGP9.5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | UCHL1/PGP9.5 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16843524
|
Proteintech
14730-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16833524
|
Proteintech
14730-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PGP9.5 (Protein gene product 9.5, UCH-L1, PARK5) is a neuron specific protein, structurally and immunologically distinct from neuron specific enolase. PGP9.5 has a molecular weight of 27 kDa and was first identified by high resolution two dimensional PAGE. PGP9.5 is a member of ubiquitin carboxyl-terminal hydrolase family 1 (peptidase family C12) with a ubiquitin carboxyl-terminal hydrolase domain. PGP9.5 is well known for having ubiquitin hydrolase and ligase activities that hydrolyzes small C-terminal adducts of ubiquitin to generate ubiquitin monomers. PGP9.5 is present in neurons and nerve fibers at all levels of the central and peripheral nervous system, in neuroendocrine cells, in segments of the renal tubules, in spermatogonia and Leydig cells of the testis, in ova and in some cells of both the pregt and non-pregt corpus luteum. Over expression of PGP9.5 leads to non-small cell lung cancer while decreased expression leads to Huntington disease and Alzheimer disease. Since PGP9.5 is present in cellular inclusions, it can be a useful as a neuronal marker and in the studies of neurodegenerative disorders such as with Parkinson disease.Specifications
UCHL1/PGP9.5 | |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P09936, Q00981, Q9R0P9 | |
22223, 29545, 7345 | |
UCHL1/PGP9.5 Fusion Protein Ag6490 | |
Primary | |
-20°C | |
UCHL1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, Ubiquitin thioesterase L1, Uch L1, UCHL1, UCH-L1 | |
UCHL1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |