missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCN2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | UCN2 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839763
|
Proteintech
12198-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829763
|
Proteintech
12198-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Two major regulatory peptides were originally isolated from fish urophysial extracts, Urotensin I and II. In both frog and human, the Urotensin II sequence is located at the carboxy-terminal position of the precursor. Human Urotensin II is composed of only 11 amino acid residues, while fish and frog Urotensin II possess 12 and 13 amino acid residues, respectively. The cyclic region of Urotensin II, which is responsible for the biological activity of the peptide, has been fully conserved from fish to human. However, several substitutions have occurred in the amino-terminal region of the molecule. A human G protein-coupled receptor, GPR14, is the Urotensin II receptor. Human Urotensin II is found within both vascular and cardiac tissue, including coronary atheroma, and effectively constricts isolated arteries from nonhuman primates. Urotensin II may act as an autocrine and/or paracrine hormone rather than as a circulating hormone, by playing an important role in the development of ventricular hypertrophy induced by chronic hypoxia.Specifications
UCN2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q96RP3 | |
90226 | |
UCN2 Fusion Protein Ag2836 | |
Primary | |
-20°C | |
UCN2 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
SRP, Stresscopin related peptide, UCN II, UCN2, UCNI, UR, urocortin 2, Urocortin II, Urocortin related peptide, URP | |
UCN2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |