missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25223-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mitochondrial uncoupling proteins (UCP) are members of the larger family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is widely expressed in many tissues with the greatest abundance in brain and testis. The gene product has an N-terminal hydrophobic domain that is not present in other UCPs. Two splice variants have been found for this gene.Specifications
UCP5 | |
Polyclonal | |
Unconjugated | |
SLC25A14 | |
BMCP 1, BMCP1, SLC25A14, UCP 5, UCP5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9016 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95258 | |
SLC25A14 | |
UCP5 Fusion Protein Ag18661 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |