missing translation for 'onlineSavingsMsg'
Learn More

UGCG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12869-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16850814

  • 1656.73 SEK / 20µL
Estimated Shipment: 29-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May serve as a "flippase" as well as a glucosyltransferase that transfers glucose to ceramide. It Catalyzes; UDP-glucose + Nacylsphingosine = UDP + D-glucosyl-N-acylsphingosine. Ceramide Glycosyltransferase is the first step in the Glycosphingolipid synthesis; first glycosylation step. Glucosylceramide synthase (GlcT) and lactosylceramide synthase (GalT) are key enzymes for the synthesis of major glycosphingolipids of vertebrates.
Specifications

Specifications

UGCG
Polyclonal
Unconjugated
UGCG
Ceramide glucosyltransferase, GCS, GLCT 1, Glucosylceramide synthase, UGCG
Rabbit
Antigen Affinity Chromatography
RUO
22234, 7357, 83626
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O88693, Q16739, Q9R0E0
UGCG
UGCG Fusion Protein Ag3530
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers