missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10391-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The enzyme encoded by this gene is an important intermediary in mammalian carbohydrate interconversions. It transfers a glucose moiety from glucose-1-phosphate to MgUTP and forms UDP-glucose and MgPPi. In liver and muscle tissue, UDP-glucose is a direct precursor of glycogen; in lactating mammary gland it is converted to UDP-galactose which is then converted to lactose. The eukaryotic enzyme has no significant sequence similarity to the prokaryotic enzyme. Two transcript variants encoding different isoforms have been found for this gene.Specifications
UGP2 | |
Polyclonal | |
Unconjugated | |
UGP2 | |
pHC379, UDP glucose pyrophosphorylase, UDPG, UDPGP, UDPGP2, UGP1, UGP2, UGPase, UGPP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216558, 289827, 7360 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16851, Q91ZJ5 | |
Ugp2 | |
UGP2 Fusion Protein Ag0590 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |