missing translation for 'onlineSavingsMsg'
Learn More

UGT3A2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21366-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880965

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

UGT3A2
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3SY77
UDPGT 3A2, UGT3A2, UNQ842/PRO1780
Rabbit
IgG
150 μL
-20°C
Primary
167127
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UGT3A2
Liquid
UGT3A2
UGT3A2 Fusion Protein Ag15835
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers