Läs mer
Tocris Bioscience™ UK 370106
Beskrivning
Highly selective MMP-3 and MMP-12 inhibitor (IC50 values are 0.023, 0.042, 1.75, 2.3, 5.8, 30.4, 34.2 and 66.9 μM at MMP 3, 12, 8, 13, 7, 9, 2 and 14 respectively.) Inhibits fibronectin cleavage (IC50 = 320 nM) and has little effect on keratinocyte migration in vitro. Substantially inhibits MMP-3 in an ex vivo model of chronic dermal ulcers.
Specifikationer
Specifikationer
| Kemiskt namn eller material | UK 370106 |
| CAS | 230961-21-4 |
| Kvantitet | 10 mg |
| Molekylformel | C35H44N2O5 |
| Formel vikt | 572.73 |
| Renhet | >95% |
| Molekylvikt (g/mol) | 572.746 |