missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ UL49 Polyclonal Antibody

Beskrivning
UL49 Polyclonal Antibody for Western Blot, ELISA
UL49 is a gene that is found in the genome of certain viruses, specifically in the family Herpesviridae. UL49 is a nonessential gene, meaning that it is not essential for the replication and survival of the virus, but it may play a role in viral pathogenesis and host immune evasion. The specific function of UL49 can vary depending on the virus in which it is found. For example, in herpes simplex virus type 1 (HSV-1), UL49 encodes a glycoprotein that is involved in the viral entry process. In other herpesviruses, the exact function of UL49 may be different.
Specifikationer
Specifikationer
| Antigen | UL49 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 2.851 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene Alias | UL49; Uncharacterized protein UL49 |
| Värd art | Rabbit |
| Immunogen | Recombinant Human cytomegalovirus (strain AD169) (HHV-5) (HCMV) Uncharacterized protein UL49 (354-570aa). |
| Reningsmetod | Protein G |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?