Learn More
ULBP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564083
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AHSLWYNFTI IHLPRHGQQW CEVQSQVDQK NFLSYDCGSD KVLSMGHLEE QLYATDAWGK QLEM Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 27%.
UL16-binding protein 3 (ULBP3) is an extended member of the MHC class I superfamily. It does not contain the alpha 3 domain and lacks a transmembrane domain. ULBP3 binds and activates the KLRK1/NKG2D receptor and mediates natural killer cell cytotoxicity.Specifications
ULBP3 | |
Polyclonal | |
Unconjugated | |
ULBP3 | |
ALCAN-gamma; N2DL3; N2DL-3; NKG2D ligand 3; NKG2D ligand3; NKG2DL3; RAET1N; Retinoic acid early transcript 1N; UL16 binding protein 3; UL16-binding protein 3; ULBP; ULBP 3; ULBP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79465 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZM4 | |
ULBP3 | |
Recombinant protein corresponding to Human ULBP3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only