Läs mer
Ulipristal Acetate, TRC
Produktkod. 30004272
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004272 10 mg
30003314 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004272 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU70081010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126784-99-4
Molekylformel C30 H37 N O4
Molekylvikt (g/mol) 475.62
Synonym 19-Norpregna-4,9-diene-3,20-dione, 17-(acetyloxy)-11-[4-(dimethylamino)phenyl]-, (11β)-, (11β)-17-(Acetyloxy)-11-[4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, 17-Acetoxy-11β-[(4-(N,N-dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, 17-Acetoxy-11β-[(4-(dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, 17-Acetyloxy-11β-[(4-(N,N-dimethylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, 17α-Acetoxy-11β-(4-dimethylaminophenyl)-19-norpregna-4,9-dien-3,20-dione, 17α-Acetoxy-11β-[4-N,N-dimethylaminophenyl]-19-norpregna-4,9-diene-3,20-dione, CBD(VA) 2914, CDB 2914, EllaOne, Esmya, HRP 2000, RTI 3021-012, RU 44675, Ulipristal acetate, VA 2914
IUPAC-namn [(8S,11R,13S,14S,17R)-17-acetyl-11-[4-(dimethylamino)phenyl]-13-methyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
LEDER CN(C)c1ccc(cc1)[C@H]2C[C@@]3(C)[C@@H](CC[C@]3(OC(=O)C)C(=O)C)[C@@H]4CCC5=CC(=O)CCC5=C24
Specifikationer
Kemiskt namn eller material Ulipristal Acetate
Kvantitet 10 mg
InChI formel InChI=1S/C30H37NO4/c1-18(32)30(35-19(2)33)15-14-27-25-12-8-21-16-23(34)11-13-24(21)28(25)26(17-29(27,30)3)20-6-9-22(10-7-20)31(4)5/h6-7,9-10,16,25-27H,8,11-15,17H2,1-5H3/t25-,26+,27-,29-,30-/m0/s1
Formel vikt 475.2723