Läs mer
ULK-101, MedChemExpress
Produktkod. 16477587
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16477587 5 mg 5mg
16474838 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16477587 Leverantör Medchem Express Leverantörsnummer HY114490
MedChemExpress ULK-101 is a potent and selective ULK1 inhibitor, with IC50 values of 1.6 nM and 30 nM for ULK1 and ULK2, respectively. ULK-101 suppresses autophagy and sensitizes cancer cells to nutrient stress.
ULK-101 is a potent and selective ULK1 inhibitor, with IC50 values of 1.6 nM and 30 nM for ULK1 and ULK2, respectively. ULK-101 suppresses autophagy and sensitizes cancer cells to nutrient stress.
Specifikationer
Kemiskt namn eller material ULK-101
CAS 2443816-45-1
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C22H16F4N4OS
Kvantitet 5 mg
Löslighetsinformation DMSO : 83.33 mg/mL (180.98 mM; Need ultrasonic)
LEDER FC1=CC=C(C(C=N2)=CN3C2=C(C4=CSC(C(N[C@@H](C5CC5)C(F)(F)F)=O)=C4)C=N3)C=C1
Molekylvikt (g/mol) 460.45
Formel vikt 460.45
Procent renhet 99.3%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death