Läs mer
MedChemExpress Ultra High Sensitivity ECL Kit
Produktkod. 16442336
Encompass_Preferred
Change view
Kvantitet:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16442336 500 ml
16468946 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16442336 Leverantör MedChemExpress Leverantörsnummer HYK1005
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
MedChemExpress MCE Ultra High Sensitivity ECL Kit is an ultra-sensitive, luminol-based enhanced chemiluminescent substrate for detecting horseradish peroxidase (HRP) labeled antibodies on western blots.
Specifikationer
Produkttyp Kits
Kvantitet 500 ml
Innehåll och lagring Stored at 4°C, and is stable for 1 year. Protect from light.
Frakt skick Room temperature
Mål VD/VDR