Learn More
BRAND™ Ultra Low Retention Filter Tips
Significantly reduce sample loss and provide substantially higher reproducibility when working with critical media.
496.00 SEK - 563.00 SEK
Specifications
Description | Ultra Low Retention |
---|---|
Filtered | Filtered |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Certifications/Compliance | CE-IVD, DE-M |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15184977
|
BRAND™
732804 |
0.5 to 10 μL |
553.00 SEK
Pack of 480 |
In Stock
Log in to see stock. |
|||||
15104987
|
BRAND™
732808 |
2 to 20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15114987
|
BRAND™
732810 |
5 to 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15124987
|
BRAND™
732812 |
5 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15154987
|
BRAND™
732814 |
50 to 1000 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Cleanroom quality for contamination-free analyses
- Perfect fit for the Transferpette™ micropipette and many other pipettes
- Polypropylene construction with polyethylene filter
- TipBox packaging
- Non-sterile
- Autoclavable at 121°C
- DIN EN 285 and CE-marked according to IVD-Directive 98/79 EC
- DNA-, RNase-, endotoxin-free
Specifications
Ultra Low Retention | |
Non-sterile | |
CE-IVD, DE-M | |
480/PK |
Filtered | |
Autoclavable | |
TipBox | |
Polypropylene, Polyethylene (Filter) |