Läs mer
Systec™ Ultracooler Heat Exchanger H-210 to H-1580 Autoclaves
För användning med (utrustning):
H-210 to H-1580 Autoclave
Förpackningsstorlek:
Varje
Beskrivning
- In conjunction with optional mantle cooling with cooling water and support pressure, it is possible to significantly reduce the recooling time and the entire sterilization process via integration of additional ultracooler heat exchanger
- The ultracooler is placed in the door of the sterilization chamber near the radial ventilator so that the usable space in the autoclave is not reduced.
- Reduction of cooling time by up to 90%
- Depending on the load, cooling times between 15 and 60 minutes can be achieved.
Specifikationer
Specifikationer
| Beskrivning | Reduces cooling time by up to 90% |
| För användning med (applikation) | Accelerated Autoclave Cooling |
| För användning med (utrustning) | H-210 to H-1580 Autoclave |
| Produkttyp | Ultracooler |