Learn More
Cole-Parmer™ Ultrapure Application Filter Cartridges
Tolerant to an accumulated exposure of 10 hours. Filters may also be sanitized with compatible chemical agents.
1785.00 SEK - 7650.00 SEK
Specifications
For Use With (Application) | Ultrapure Applications |
---|---|
Type | Ultrapure Application Filter Cartridge |
Length (English) | 10 in. |
Length (Metric) | 25.4 cm |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11767203
|
Cole-Parmer™
06479-36 |
Double open end polypropylene filter cartridge, 1.2µm |
1890.00 SEK
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
11706119
|
Cole-Parmer™
06479-40 |
Double open end polypropylene filter cartridge, 5µm |
1785.00 SEK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11716119
|
Cole-Parmer™
06479-48 |
Double open end PTFE filter cartridge, 0.1µm |
7650.00 SEK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11726119
|
Cole-Parmer™
06479-52 |
Double open end PTFE filter cartridge, 0.2µm |
6955.00 SEK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
Description
- These cartridges are autoclavable to 250°F 121°C) and in-line sterilizable to 257°F (125°C) for up to 30-minute cycles
- Tolerant to an accumulated exposure of 10 hours
- Filters may also be sanitized with compatible chemical agents
- Suitable to 180°F (82°C) at 10psid
- The cartridges comply with U.S. CFR Title 21 guidelines for repeated food contact, comply with USP Class VI, 121°C Plastics guidelines
- Pass the MEM Elution Cytotoxicity Test
- Aqueous extracts contain less than 0.25 EU/mL
- Made of resin-bonded borosilicate glass microfibers that do not leach flavor-altering substances
- Meet NSF Standard 53 for the reduction of cysts
- Ideal as prefilters to protect membrane filter/RO systems
- Multi-Layer Nylon Cartridges.
- These consist of two pleated nylon membranes with the top acting as a prefilter to increase cartridge life and efficiency.
- Glass Microfiber Cartridges
- Made of resin-bonded borosilicate glass microfibers that do not leach flavor-altering substances. Meet NSF Standard 53 for the reduction of cysts. Ideal as prefilters to protect membrane filter/RO systems.
- Polypropylene (PP) Cartridges
- Thermally bonded high-purity PP fiber. Economical for a chemically inert media with high dirtloading capacity. Suitable for air/gas filtration; FDA-acceptable. Hydrophobic; prewet before liquid filtration.
- Pleated PTFE Cartridges
- Clean room-constructed using thermal welding instead of adhesives or additives. Ideal for the more aggressive liquid or air/gas filtration applications. Hydrophobic; prewet before liquid filtration.
- Semiconductor Cartridges
- Single-layer PES membrane over PP core. Each is flushed with high-purity water to leach less than 5 ppb TOC and less than 5 ppb trace metals. Ideal for any highpurity chemical or DI water application.
Specifications
Ultrapure Applications | |
Ultrapure Application Filter Cartridge | |
10 in. | |
25.4 cm |