Läs mer
Umckalin, TRC
Produktkod. 30455990
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455990 10 mg
30450502 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30455990 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU70095010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 43053-62-9
Molekylformel C11 H10 O5
Molekylvikt (g/mol) 222.19
Synonym 2H-1-Benzopyran-2-one, 7-hydroxy-5,6-dimethoxy-, 7-Hydroxy-5,6-dimethoxy-2H-1-benzopyran-2-one, Umckalin
IUPAC-namn 7-hydroxy-5,6-dimethoxychromen-2-one
LEDER COc1c(O)cc2OC(=O)C=Cc2c1OC
Specifikationer
Kemiskt namn eller material Umckalin
Kvantitet 10 mg
InChI formel InChI=1S/C11H10O5/c1-14-10-6-3-4-9(13)16-8(6)5-7(12)11(10)15-2/h3-5,12H,1-2H3
Formel vikt 222.0528