missing translation for 'onlineSavingsMsg'
Läs mer

UNC 0631, MedChemExpress

Produktkod. 16492017
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16492017 50 mg 50mg
16485868 10 mg -
16473386 25 mg -
16408527 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16492017 Leverantör Medchem Express Leverantörsnummer HY13808

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.

UNC 0631 is a potent histone methyltransferase G9a inhibitor with an IC50 of 4 nM. UNC 0631 potently reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 25 nM.

Specifikationer

Kemiskt namn eller material UNC 0631
CAS 1320288-19-4
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C37H61N7O2
Kvantitet 50 mg
Löslighetsinformation DMF : 150 mg/mL (235.88 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (157.25 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (26.21 mM; Need ultrasonic)
LEDER COC1=CC2=C(NC3CCN(CC4CCCCC4)CC3)N=C(N5CCN(C(C)C)CCC5)N=C2C=C1OCCCN6CCCCC6
Molekylvikt (g/mol) 635.93
Formel vikt 635.93
Procent renhet 99.35%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death
Visa mer Visa mindre

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.