Läs mer
UNC0379 TFA, MedChemExpress
Produktkod. 16427827
Encompass_Preferred
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
16427827 5 mg
1 options
Produktkod. 16427827 Leverantör Medchem Express Leverantörsnummer HY12335A
MedChemExpress UNC0379 TFA is a selective, substrate-competitive inhibitor of lysine methyltransferase SETD8 (KMT5A) with an IC50 of 7.3 μM, KD value of 18.3 μM. UNC0379 TFA can be used in the research of inflammation and cancers, such as pulmonary fibrosis, ovarian cancer, neuroblastoma.
UNC0379 TFA is a selective, substrate-competitive inhibitor of lysine methyltransferase SETD8 (KMT5A) with an IC50 of 7.3 μM, KD value of 18.3 μM. UNC0379 TFA can be used in the research of inflammation and cancers, such as pulmonary fibrosis, ovarian cancer, neuroblastoma.
Specifikationer
Kemiskt namn eller material UNC0379 TFA
CAS 1620401-83-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C25H36F3N5O4
Kvantitet 5 mg
Löslighetsinformation H2O : 100 mg/mL (189.54 mM; Need ultrasonic)
LEDER COC1=C(OC)C=C2C(N=C(N3CCCC3)N=C2NCCCCCN4CCCC4)=C1.O=C(O)C(F)(F)F
Molekylvikt (g/mol) 527.58
Formel vikt 527.58
Procent renhet 99.46%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death