Läs mer
UNC1999, MedChemExpress
Produktkod. 16496856
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16496856 100 mg 100mg
16469267 5 mg -
16432797 50 mg -
16416718 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496856 Leverantör Medchem Express Leverantörsnummer HY15646
MedChemExpress UNC1999 is a SAM-competitive, potent and selective inhibitor of EZH2/1 with IC50s of <10 nM and 45 nM, repectively.
UNC1999 is a SAM-competitive, potent and selective inhibitor of EZH2/1 with IC50s of <10 nM and 45 nM, repectively.
Specifikationer
Kemiskt namn eller material UNC1999
CAS 1431612-23-5
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C33H43N7O2
Kvantitet 100 mg
Löslighetsinformation DMSO : 100 mg/mL (175.52 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble)
LEDER O=C1C(CNC(C2=C(C=NN3C(C)C)C3=CC(C4=CN=C(N5CCN(C(C)C)CC5)C=C4)=C2)=O)=C(CCC)C=C(C)N1
Molekylvikt (g/mol) 569.74
Formel vikt 569.74
Procent renhet 98.86%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death