Learn More
UNC84A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552548
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QLLPTVEHLQ LELDQLKSEL SSWRHVKTGC ETVDAVQERV DVQVREMVKL LFSEDQQGGS LEQLLQRFSS QFVSKGDLQT MLRDLQLQIL RNVTHHVSVT KQLPTSEAVV SAVSEAGASG ITEAQARAIV NS Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 58%.
UNC84A Polyclonal antibody specifically detects UNC84A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
UNC84A | |
Polyclonal | |
Unconjugated | |
SUN1 | |
4632417G13Rik; 5730434D03Rik; KIAA0810; mKIAA0810; Protein unc-84 homolog A; Sad1 and UNC84 domain containing 1; Sad1 unc-84 domain protein 1; sad1/unc-84 protein-like 1; SUN domain-containing protein 1; Sun1; unc-84 homolog A; Unc84a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23353 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94901 | |
SUN1 | |
Recombinant protein corresponding to Human UNC84A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only