missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecanoic-d21 Acid, TRC
Specifications
Specifications
| Kemiskt namn eller material | Undecanoic-D21 Acid |
| CAS | 60658-40-4 |
| Molekylformel | C11HD21O2 |
| Kvantitet | 10 mg |
| Synonym | Undecanoic-d21 acid (9CI), Undecanoic acid D21, Undecanoic acid-d21, Perdeuteroundecanoic acid |
| InChI formel | InChI=1S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| LEDER | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O |
| IUPAC-namn | undecanoic-d21 acid |
| Molekylvikt (g/mol) | 207.42 |
| Formel vikt | 207.29 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?