Learn More
Abeba™ UNI6 36760 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand:  Abeba™ 36760/45
Additional Details : Weight : 0.98000kg
 View more versions of this product
Product Code. 15670548
  • 1251.93 SEK / 1 pair
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Functional leather with honeycomb pattern
  • White
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Mater-repellent
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Leather
Slip-resistant
36760
White
45
Polyurethane