missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Unipur Carbon FT Safety Gloves

missing translation for 'orderingAttributeHoverText'
Size:
6
7
8
9
10
11
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Sensitive, anti-static safety gloves feature a thin elastomer coating on the fingertips for precision work with electronics parts.

  • Suited for precision mechanical work in antistatic environments
  • Thin elastomer coating on the fingertips for enhanced grip and tactility
  • Polyamide/carbon liner provides a close fit for increased dexterity
  • Fulfils the requirements of DIN EN 16350:2014
Certifications/Compliance (Europe) DIN EN 16350:2014
Coated Area Fingertips
Applications Industrial
Coating Material Elastomer
Color Gray
Material Polyamide, Carbon
Size 8
EN388 Standard 2131X
For Use With (Application) Electronics, Precision Assembly
Gender Unisex
Glove Cuff Style Knit
Series Unipur Carbon FT
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'