Läs mer
IBA Solutions For Life Science™ Universal magnet plate flat
Brand:  IBA Solutions For Life Science™ 7-2011-000
Produktkod. 12763084
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
  • Universal magnet plate (80mm x 130mm) suitable for Magnet Assisted Transfection of cells grown in culture plates, e.g., 6 well, 12 well, 24 well, 48 well or 96 well plates, T75 flasks, 60mm dishes etc.
  • Universal magnet plate (260mm x 260mm) suitable for Magnet Assisted Transfection of cells grown in culture plates, e.g., 6 well, 12 well, 24 well, 48 well or 96 well plates, T75 flasks, T175 flasks, 60mm dishes, 100mm dishes etc.
  • For 96 well culture plates a special 96 magnet bar plate (80mm x 120mm) optimised for transfection in 96 individual wells
  • Compatible with culture plates from the most common suppliers
  • Can easily be decontaminated with 70% ethanol
  • Can be used within incubators and robots