Learn More
Thermo Scientific™ MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Brand: Thermo Scientific™ 30115BI
13153.23 SEK valid until 2024-12-31
Use promo code "21970" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Shaker Platform | |
30 x 18 in. (76 x 45.7 cm) | |
76 cm | |
45.7 cm | |
MaxQ 3000, 5000 |
Certificate of Analysis (COA), Certificate of Conformance (COC) | |
30 in. | |
18 in. | |
Universal Platform without Clamps, 18 x 30 in. |