Learn More
Idex Male-Threaded Nuts
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
Brand: Kinesis™ U-400X
Additional Details : Weight : 0.25000kg
Description
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
- Eliminates tubing twist that can loosen connections
- Resistant to loosening through vibration
Specifications
0.25 in. | |
1/16 in. O.D. Tubing | |
Stainless Steel |
0.4 in. | |
0.0625 in. | |
Nut |