Learn More
Kinesis™ Precolumn Inline Filters
Protect larger columns and injections from harmful particulates
Brand: Kinesis™ 02010-35
Additional Details : Weight : 0.25000kg
Description
- Ideal for higher-flow systems
- Interchangeable frits make protection more economical
- Compatible with SFC and HPLC
- Available to fit a variety of tubing sizes
Specifications
0.5 μm | |
PEEK | |
Precolumn Inline Filter | |
1.3 uL |