Läs mer
UPF-648, MedChemExpress
Produktkod. 16466668
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16466668 10 mg
16449227 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16466668 Leverantör Medchem Express Leverantörsnummer HY15600
MedChemExpress UPF-648 is a potent kynurenine 3-monooxygenase (KMO) inhibitor; exhibits highly active at 1 uM (81 ± 10% KMO inhibition); ineffective at blocking KAT activity.
UPF-648 is a potent kynurenine 3-monooxygenase (KMO) inhibitor
Specifikationer
Kemiskt namn eller material UPF-648
Fysisk form Solid
CAS 213400-34-1
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C11H8Cl2O3
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation Ethanol : ≥ 50 mg/mL (192.98 mM)
LEDER O=C([C@@H]1[C@@H](C(C2=CC=C(Cl)C(Cl)=C2)=O)C1)O
Molekylvikt (g/mol) 259.09
Formel vikt 259.09
Procent renhet 99.86%