missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563120
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TGANAEKAES HNDCPVRLLN PNIAKMKEDI LYHFNLTTSR HNFP Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 63%.
UPP1 catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate (PubMed:7488099). The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis.Specifications
UPP1 | |
Polyclonal | |
Unconjugated | |
UPP1 | |
AI325217; im:6911242; UDRPASE; UP; UPase; UPase 1; UPP; Upp1; urdPase 1; uridine phosphorylase 1; zgc:110755 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7378 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16831 | |
UPP1 | |
Recombinant protein corresponding to Human UPP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only