Läs mer
Tocris Bioscience™ uPSEM 792 hydrochloride
Beskrivning
uPSEM 792 hydrochloride is a ultrapotent PSEM (uPSEM) agonist for PSAM4-GlyR and PSAM4-5HT3 (Ki = 0.7 nM for PSAM4-GlyR and <10 nM for PSAM4-5HT3). Exhibits >10,000-fold agonist selectivity for PSAM4-GlyR over α7-GlyR, α7-5HT3, and 5HT3-R, and 230-fold selectivity over α4β2 nAChR. Also weak partial agonist (∽10 %) at α4β2 nAChR. Retains the potency of varenicline (Cat.No. 3754) for PSAM4-GlyR with enhanced chemogenetic selectivity. Does not act as a substrate for P-glycoprotein pumps. Silences neurons in vivo. Brain-penetrant.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-Methyl-7,8,9,10-tetrahydro-1 H-6,10-methanoazepino[4,5-g]quinoxalin-2(6 H)-one hydrochloride |
| CAS | 2341841-08-3 |
| Kvantitet | 25 mg |
| Mål | Pharmacologically Selective Effector Molecules/PSEM |
| Molekylformel | C14H15N3O.HCl |
| Formel vikt | 277.75 |
| Renhet | 98% |