missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | UQCC |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826005
|
Proteintech
28114-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816005
|
Proteintech
28114-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.Specifications
UQCC | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9NVA1 | |
55245 | |
UQCC Fusion Protein Ag27123 | |
Primary | |
-20°C | |
UQCC1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BFZB, BZFB, C20orf44, CBP3, UQCC | |
UQCC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |