Läs mer
Uracil-15N2, TRC
Produktkod. 30449266
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449266 250 mg
30452533 500 mg
30449056 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30449266 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU801105250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5522-55-4
Molekylformel C4H415N2O2
Molekylvikt (g/mol) 114.07
Synonym 15N2-Uracil, 2,4(1H,3H)-Pyrimidinedione-1,3-15N2
IUPAC-namn (1,3-^{15}N_{2})1H-pyrimidine-2,4-dione
LEDER O=C1[15NH]C=CC(=O)[15NH]1
Specifikationer
Kemiskt namn eller material Uracil-15N2
Kvantitet 250 mg
InChI formel InChI=1S/C4H4N2O2/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)/i5+1,6+1
Formel vikt 114.0213