missing translation for 'onlineSavingsMsg'
Learn More

Uracil, 99.75%, MP Biomedicals™

Catalog Number p-4404572
Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy
66-22-8
C4H4N2O2
112.09
MFCD00006016
ISAKRJDGNUQOIC-UHFFFAOYSA-N
uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
1174
CHEBI:17568
1,2,3,4-tetrahydropyrimidine-2,4-dione
O=C1NC=CC(=O)N1
This item is not returnable. View return policy
66-22-8
C4H4N2O2
112.09
MFCD00006016
ISAKRJDGNUQOIC-UHFFFAOYSA-N
uracil, 2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 2,4-pyrimidinediol, 2,4-dioxopyrimidine, pyrimidine-2,4 1h,3h-dione, pirod, 2,4-pyrimidinedione, pyrod, hybar x
1174
CHEBI:17568
1,2,3,4-tetrahydropyrimidine-2,4-dione
O=C1NC=CC(=O)N1
Color White
Assay Percent Range 99.75%
Quantity 25 g
Formula Weight 112.09
Physical Form Crystalline Powder
Chemical Name or Material Uracil
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'