Learn More
Corning™ PureCoat™ Carboxyl Rectangular Straight Neck Cell Culture Flask with Vented Cap
Improved cell attachment, faster cell proliferation and enhance recovery post-thaw over standard TC surfaces
Brand:  Corning™ 356780
Additional Details : Weight : 6.39000kg
Product Code. 11957151
  • 5184.49 SEK / Pack of 40
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
Straight
Rectangular
Carboxyl
Yes
Special Offers
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.