Läs mer
Uridine-2-13C, CDN
Produktkod. 30319078
Change view
Kvantitet:
0.05 g
Produktkod. Kvantitet
30319078 0.05 g
1 options
Produktkod. 30319078 Leverantör CDN Leverantörsnummer CDNC33920.05G
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Uridine-2-13 C
CAS 35803-42-0
Molekylformel C8(13 C)H12N2O6
Kvantitet 0.05 g
Synonym Uridine-2-13 C (9 CI), [2-13 C]Uridine, 1-β-D-Ribofuranosyl-2,4(1 H,3 H)-pyrimidinedione-2-13 C, 1-β-D-Ribofuranosyluracil-2-13 C
InChI formel InChI=1 S/C9H12N2O6/c12-3-4-6(14)7(15)8(17-4)11-2-1-5(13)10-9(11)16/h1-2,4,6-8,12,14-15 H,3H2,(H,10,13,16)/t4-,6-,7-,8-/m1/s1/i9+1
LEDER O[C@@H]1 [C@H](O)[C@@H](CO)O[C@H]1 N(C=C2)[13 C](NC2=O)=O
IUPAC-namn 1-((2 R,3 R,4 S,5 R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1 H,3 H)-dione-2-13 C
Molekylvikt (g/mol) 245.2
Formel vikt 245.07 g/mol
Procent renhet 99 atom % 13 C, min 98% Chemical Purity