Läs mer
Uridine 5’-Diphospho-N-acetylglucosamine Disodium Salt, TRC
Produktkod. 30456819
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30456819 10 mg
1 options
Produktkod. 30456819 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU82992510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Uridine 5'-Diphospho-N-acetylglucosamine Disodium Salt
CAS 91183-98-1
Molekylformel C17 H25 N3 O17 P2 . 2 Na
Kvantitet 10 mg
Synonym Uridine 5'-(trihydrogen diphosphate)','P'-[2-(acetylamino)-2-deoxy-α-D-glucopyranosyl] ester','sodium salt (1:2), Uridine 5'-(trihydrogen diphosphate)','P'-[2-(acetylamino)-2-deoxy-α-D-glucopyranosyl] ester','disodium salt (9CI), Uridine 5'-(trihydrogen diphosphate) P'-[2-(acetylamino)-2-deoxy-α-D-glucopyranosyl] ester disodium salt, Uridine 5'-diphospho-N-acetylglucosamine disodium salt, UDP-N-acetyl-D-glucosamine disodium salt, UDP-N-acetylglucosamine disodium salt, UDP-GlcNAc disodium salt, UDP-α-D-N-acetylglucosamine disodium salt, Uridine 5'-(2-acetamido-2-deoxy-α-D-glucosyl pyrophosphate) disodium salt
InChI formel InChI=1S/C17H27N3O17P2.2Na/c1-6(22)18-10-13(26)11(24)7(4-21)35-16(10)36-39(31,32)37-38(29,30)33-5-8-12(25)14(27)15(34-8)20-3-2-9(23)19-17(20)28;;/h2-3,7-8,10-16,21,24-27H,4-5H2,1H3,(H,18,22)(H,29,30)(H,31,32)(H,19,23,28);;/q;2*+1/p-2/t7-,8-,10-,11-,12-,13-,14-,15-,16-;;/m1../s1
LEDER [Na+].[Na+].CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1OP(=O)([O-])OP(=O)([O-])OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O
IUPAC-namn disodium;[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl] [[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate
Molekylvikt (g/mol) 651.32
Formel vikt 651.0455