Läs mer
Uridine-5'-triphosphate Trisodium Salt Dihydrate (>90%), TRC
Produktkod. 30453358
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30453358 1 g
1 options
Produktkod. 30453358 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU8300481G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Uridine-5'-triphosphate Trisodium Salt Dihydrate
CAS 116295-90-0
Molekylformel C9 H12 N2 Na3 O15 P3 . 2(H2 O)
Kvantitet 1 g
Synonym Uridine 5'-(tetrahydrogen triphosphate) sodium salt hydrate (1:3:2), Uridine 5'-(tetrahydrogen triphosphate) trisodium salt dihydrate (9CI), UTP trisodium dihydrate
InChI formel InChI=1S/C9H15N2O15P3.3Na.2H2O/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H,10,12,15)(H2,16,17,18);;;;2*1H2/q;3*+1;;/p-3/t4-,6-,7-,8-;;;;;/m1...../s1
LEDER O.O.[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)(O)OP(=O)([O-])OP(=O)([O-])[O-])N2C=CC(=NC2=O)O
IUPAC-namn trisodium;[[[(2R,3S,4R,5R)-3,4-dihydroxy-5-(4-hydroxy-2-oxo-pyrimidin-1-yl)tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl]oxy-oxido-phosphoryl] phosphate;dihydrate
Molekylvikt (g/mol) 586.12
Formel vikt 585.9355