Learn More
UROD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555630
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ALVPYLVGQV VAGAQALQLF ESHAGHLGPQ LFNKFALPYI RDVAKQVKAR LREAGLAPVP MIIFAKDGHF Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
UROD Polyclonal antibody specifically detects UROD in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
UROD | |
Polyclonal | |
Unconjugated | |
Urod | |
AI323803; PCT; porphyrinogen carboxylase; porphyrinogen carboxy-lyase; UPD; URO D; UROD; Uro-d; Uroporphyrinogen decarboxylase; uroporphyrinogen III decarboxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7389 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P06132 | |
Urod | |
Recombinant protein corresponding to Human UROD | |
100 μL | |
Primary | |
Human | |
Antibody | |