missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UROS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14913-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene catalyzes the fourth step of porphyrin biosynthesis in the heme biosynthetic pathway. Defects in this gene cause congenital erythropoietic porphyria (Gunther′s disease).Specifications
UROS | |
Polyclonal | |
Unconjugated | |
UROS | |
UROIIIS, uroporphyrinogen III synthase, UROS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22276, 309070, 7390 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10746, P51163 | |
Uros | |
UROS Fusion Protein Ag6711 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |