missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Urotensin II Receptor Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16620855 Handla allt Thermo Scientific Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
16620855 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16620855 Leverantör Invitrogen™ Leverantörsnummer PA5104401
  • ONLINE EXCLUSIVE PRICE
    5330.00 SEK
    4530.50 SEK / 100µL
    Save 799.50 SEK
    15% Av
 Begränsad artikel
Beräknad leverans: 25-09-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UTS2R.

The Urotensin-II receptor has been reported to be expressed at high levels in the heart and pancreas and also in artery and smooth muscle. Low transcript levels have been identified in brain, particularly in thalamus, and superior occipital gyrus. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Urotensin II Receptor
Användningsområden Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen UTS2R
Genaccessionsnr. P49684, Q8VIH9, Q9UKP6
Gene Alias G protein-coupled receptor 14; GPR14; G-protein coupled receptor 14; G-protein coupled sensory epithelial neuropeptide-like receptor; putative G protein-coupled receptor (SENR); Senr; Uiir; Uii-r; Uii-r1a; UII-receptor; UR2R; UR-2-R; UR-II-R; urotensin 2 receptor; urotensin II receptor; Urotensin-2 receptor; UT-II; UTIIR; UTR; UTR2; Uts2r
Gensymboler UTS2R
Värd art Rabbit
Immunogen A synthesized peptide derived from human UTS2R(Accession Q9UKP6), corresponding to amino acid residues G336-R386.
Reningsmetod Affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 217369, 2837, 57305
Målarter Human, Mouse, Rat
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.