Läs mer
Ursodeoxycholic Acid-24-13C, TRC
Produktkod. 30449269
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449269 5 mg
30450202 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449269 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU8500015MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ursodeoxycholic Acid 24-13C
CAS 63296-46-8
Molekylformel C23(13C)H40O4
Kvantitet 5 mg
Synonym Cholan-24-oic-24-13C acid','3,7-dihydroxy-','(3α,5β,7β)- (9CI), Ursodeoxycholic acid 24-13C, Ursodeoxycholic-24-13C acid, 3α,7β-Dihydroxy-5β-cholan-24-oic acid-24-13C
InChI formel InChI=1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1/i21+1
LEDER C[C@@]12[C@](CC[C@]2([H])[C@H](C)CC[13C](O)=O)([H])[C@@]3([H])[C@@](CC1)([H])[C@@]4([C@](C[C@@H](CC4)O)([H])C[C@@H]3O)C
IUPAC-namn (R)-4-((3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic-1-13C acid
Molekylvikt (g/mol) 393.57
Formel vikt 393.3