Läs mer
Ursolic Acid, 98%, MP Biomedicals™
Produktkod. 11469721 Handla allt MP Biomedicals Produkter
Change view
Kvantitet:
100 mg
500 mg
Förpackningsstorlek:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11469721 100 mg 100mg
11479721 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11469721 Leverantör MP Biomedicals Leverantörsnummer 0215825380
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Kemiska identifierare
CAS 77-52-1
Molekylformel C30H48O3
Molekylvikt (g/mol) 456.71
MDL-nummer MFCD00009621
InChI-nyckel WCGUUGGRBIKTOS-GPOJBZKASA-N
Synonym 3β-Hydroxy-12-ursen-28-ic acid
PubChem CID 129316832
IUPAC-namn (1S,2R,4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
LEDER C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O
Specifikationer
Kemiskt namn eller material Ursolic Acid
Analysprocentintervall 0.98
Kvantitet 100 mg
Formel vikt 456.71
Fysisk form Pulver