Learn More
SI Analytics™ USB Cable
SI Analytics™ USB Cable is suitable for both Lab and ProLab instruments.
Brand: SI Analytics™ 285201540
Additional Details : Weight : 1.00000kg
Description
- Cable with USB connection
- For use withLab instruments
- Can be used with all ProLab meter series with USB
- Not suitable with Lab 745, 845, 850, 945
Specifications
USB |